Gene Transcripts

Gene Symbol
ABO
Entrez Gene ID
28
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Gene Aliases
A3GALNT, A3GALT1, GTB, NAGAT
Chromosome Location
Chr. 9: 133255176 - 133275214 on Build GRCh38
UniGene ID
Hs.654423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020469.2 NP_065202.2 5, 6 265
GenBank mRNA AF134412.1 - 5, 6 240
AF134413.1 - 5, 6 240
AF134414.1 - 5, 6 240
AF134415.1 - 5, 6 240
AF134416.1 - 5, 6 240
BC069595.1 - 5, 6 240 7262430
BC069814.1 - 5, 6 240
BC111575.1 - 5, 6 240 40069575
DQ139865.1 - 5, 6 240
FR691751.1 - 5, 6 240
FR691752.1 - 5, 6 240
JN791441.1 - 5, 6 240
JX398330.1 - 5, 6 240
JX398331.1 - 5, 6 240
JX398332.1 - 5, 6 240
JX398333.1 - 5, 6 240
JX519569.1 - 5, 6 240
KT282967.1 - 5, 6 240

View Full Product Details