Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000030.2 | NP_000021.1 | 7 | 829 | |
GenBank mRNA | AB292648.1 | - | 7 | 730 | |
AF348451.1 | - | 7 | 620 | ||
AK292754.1 | - | 7 | 750 | ||
AK315331.1 | - | 7 | 777 | ||
BC132819.1 | - | 7 | 826 | 40146841 | |
D13368.1 | - | 7 | 816 | ||
X53414.1 | - | 7 | 830 | ||
X56092.1 | - | 7 | 729 |