Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000033.3 | NP_000024.2 | 1 | 1273 | |
GenBank mRNA | AY421736.1 | - | 1 | 404 | |
BC015541.1 | - | 1 | 1254 | 3896490 | |
BC025358.1 | - | 1 | 1273 | 4907640 | |
Z21876.1 | - | 1 | 1260 |