Gene Transcripts

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
Chromosome Location
Chr. 7: 127588352 - 127591705 on Build GRCh38
UniGene ID
Hs.653202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001662.3 NP_001653.1 2 239
GenBank mRNA AF493884.1 - 2 92
AK297026.1 - 2 144
AK307297.1 - 2 213
AK308548.1 - 2 421
AK312107.1 - 2 144
BC003043.1 - 2 129 2822973
BC033104.1 - 2 186 4862014
BT007087.1 - 2 92
CN259625.1 - 2 256
M57567.1 - 2 146

View Full Product Details