Gene Transcripts

Gene Symbol
ARSE
Entrez Gene ID
415
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Gene Aliases
ASE, CDPX, CDPX1, CDPXR
Chromosome Location
Chr. X: 2934632 - 2968310 on Build GRCh38
UniGene ID
Hs.386975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000047.2 NP_000038.2 3 336
NM_001282628.1 NP_001269557.1 4 622
XM_005274518.2 XP_005274575.1 3 263
XM_005274519.4 XP_005274576.1 3 426
XM_011545521.1 XP_011543823.1 3 467
XM_017029525.1 XP_016885014.1 4 553
XM_017029526.1 XP_016885015.1 4 667
GenBank mRNA AK223183.1 - 3 166
AK223199.1 - 3 133
AK301004.1 - 4 453
AK313093.1 - 3 155
BP272004.1 - 3 268
BP272164.1 - 3 269
DA915010.1 - 3 338
DB073314.1 - 3 318
HX786884.1 - 3 227
X83573.1 - 3 153

View Full Product Details