Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001192.2 | NP_001183.2 | 3 | 548 | |
GenBank mRNA | AY684975.1 | - | 2 | 285 | |
BC058291.1 | - | 3 | 471 | 6736094 | |
EF152353.1 | - | 3 | 327 | ||
EF152354.1 | - | 3 | 237 | ||
EF152355.1 | - | 2 | 295 | ||
Z14954.1 | - | 3 | 396 | ||
Z29575.1 | - | 3 | 548 |