Gene Transcripts

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
Chromosome Location
Chr. 20: 57168753 - 57266651 on Build GRCh38
UniGene ID
Hs.473163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001719.2 NP_001710.1 2 995
GenBank mRNA AK291186.1 - 2 571
AK312419.1 - 2 568
BC008584.1 - 2 507 4183402
EU679366.1 - 2 379
M60316.1 - 2 562
X51801.1 - 2 588

View Full Product Details