Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001764.2 | NP_001755.1 | 5 | 1010 | |
XM_011510119.2 | XP_011508421.1 | 5 | 1352 | ||
GenBank mRNA | AI336342.1 | - | 2 | 385 | |
AK303330.1 | - | 5 | 917 | ||
AK310617.1 | - | 4 | 1282 | ||
BC069481.1 | - | 5 | 947 | 7262331 | |
BC074747.2 | - | 5 | 924 | 30915207 | |
BC104216.1 | - | 5 | 947 | 40031984 | |
BC104217.1 | - | 5 | 947 | 40031985 | |
M28826.1 | - | 5 | 947 |