Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000741.3 | NP_000732.2 | 1 | 788 | |
GenBank mRNA | AF385590.1 | - | 1 | 737 | |
AF498918.1 | - | 1 | 737 | ||
BC095546.1 | - | 1 | 738 | ||
BC119775.1 | - | 1 | 736 | ||
BC119817.1 | - | 1 | 736 | ||
BC137535.1 | - | 1 | 788 | 9021535 |