Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001859.3 | NP_001850.1 | 4 | 434 | |
GenBank mRNA | AK129613.1 | - | 4 | 376 | |
AK222866.1 | - | 4 | 376 | ||
AK292511.1 | - | 4 | 388 | ||
AL831843.1 | - | 4 | 374 | ||
BC013611.1 | - | 4 | 363 | 3881191 | |
BC061924.1 | - | 4 | 362 | 4802875 | |
U83460.1 | - | 4 | 400 |