Gene Transcripts
- Gene Symbol
- CSNK1G2
- Entrez Gene ID
- 1455
- Gene Name
- casein kinase 1 gamma 2
- Gene Aliases
- CK1g2
- Chromosome Location
- Chr. 19: 1941149 - 1981338 on Build GRCh38
- UniGene ID
- Hs.651905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319.6 |
NP_001310.3
|
3, 4 |
745 |
|
XM_005259498.1 |
XP_005259555.1
|
3, 4 |
757 |
|
XM_005259499.3 |
XP_005259556.1
|
3, 4 |
564 |
|
XM_005259500.1 |
XP_005259557.1
|
3, 4 |
757 |
|
XM_005259501.2 |
XP_005259558.1
|
3, 4 |
1131 |
|
XM_017026297.1 |
XP_016881786.1
|
3, 4 |
631 |
|
XM_017026298.1 |
XP_016881787.1
|
3, 4 |
1131 |
|
GenBank mRNA |
AB451278.1 |
- |
2, 3 |
222 |
|
AB451410.1 |
- |
2, 3 |
222 |
|
AF001177.1 |
- |
3, 4 |
1467 |
|
BC018693.2 |
- |
3, 4 |
718 |
3010717 |
BC018699.2 |
- |
3, 4 |
718 |
3010717 |
BC020972.1 |
- |
2, 3 |
275 |
3851613 |
BT009922.1 |
- |
2, 3 |
222 |
|
U89896.1 |
- |
2, 3 |
461 |
|
View Full Product Details