Gene Transcripts

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
Chromosome Location
Chr. 4: 155924118 - 155953896 on Build GRCh38
UniGene ID
Hs.75262
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001334.2 NP_001325.1 1, 2 193
GenBank mRNA AK308323.1 - 1, 2 199
AK312354.1 - 1, 2 159
BC049206.1 - 1, 2 146 5189657
BI834113.1 - 1, 2 196
X77383.1 - 1, 2 193

View Full Product Details