Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038493.1 | NP_001033582.1 | 2 | 541 | |
NM_178120.4 | NP_835221.2 | 3 | 741 | ||
GenBank mRNA | AK095266.1 | - | 1 | 2519 | |
AK290503.1 | - | 3 | 714 | ||
AY257976.1 | - | 3 | 741 | ||
BC036189.1 | - | 3 | 707 | 5311747 | |
BC053351.1 | - | 2 | 418 | 5243874 | |
KJ534824.1 | - | 3 | 543 |