Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005221.5 | NP_005212.1 | 1 | 436 | |
GenBank mRNA | AK023493.1 | - | 1 | 436 | |
AK297614.1 | - | 1 | 556 | ||
BC006226.2 | - | 1 | 422 | 3941691 | |
BT006903.1 | - | 1 | 228 | ||
DB280023.1 | - | 1 | 436 |