Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001424.5 | NP_001415.1 | 5 | 618 | |
XM_006720864.3 | XP_006720927.1 | 5 | 612 | ||
GenBank mRNA | AK313134.1 | - | 5 | 534 | |
AV657588.1 | - | 4 | 333 | ||
BC009687.1 | - | 5 | 509 | 3895094 | |
BF981157.1 | - | 5 | 600 | ||
U52100.1 | - | 4 | 453 | ||
X94770.1 | - | 4 | 400 |