Gene Transcripts
- Gene Symbol
- EPS8
- Entrez Gene ID
- 2059
- Gene Name
- epidermal growth factor receptor pathway substrate 8
- Gene Aliases
- DFNB102
- Chromosome Location
- Chr. 12: 15620141 - 15789576 on Build GRCh38
- UniGene ID
- Hs.591160
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004447.5 |
NP_004438.3
|
16, 17 |
2122 |
|
XM_011520605.2 |
XP_011518907.1
|
17, 18 |
2182 |
|
GenBank mRNA |
AK291777.1 |
- |
16, 17 |
1927 |
|
AK292931.1 |
- |
16, 17 |
1779 |
|
AK301834.1 |
- |
9, 10 |
1112 |
|
AK304860.1 |
- |
16, 17 |
2122 |
|
AK316134.1 |
- |
9, 10 |
1031 |
|
AK316239.1 |
- |
9, 10 |
1304 |
|
BC030010.2 |
- |
16, 17 |
1934 |
4820933 |
U12535.1 |
- |
16, 17 |
1894 |
|
View Full Product Details