Gene Transcripts
- Gene Symbol
- EYA3
- Entrez Gene ID
- 2140
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
- Gene Aliases
- Chromosome Location
- Chr. 1: 27970344 - 28088671 on Build GRCh38
- UniGene ID
- Hs.185774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282560.1 |
NP_001269489.1
|
7, 8 |
636 |
|
NM_001282561.1 |
NP_001269490.1
|
7, 8 |
636 |
|
NM_001282562.1 |
NP_001269491.1
|
7, 8 |
694 |
|
NM_001990.3 |
NP_001981.2
|
8, 9 |
774 |
|
NR_104214.1 |
|
8, 9 |
774 |
|
XM_006710449.3 |
XP_006710512.1
|
9, 10 |
810 |
|
XM_011540999.2 |
XP_011539301.1
|
9, 10 |
813 |
|
XM_011541001.2 |
XP_011539303.1
|
8, 9 |
771 |
|
XM_011541003.2 |
XP_011539305.1
|
8, 9 |
673 |
|
XM_011541004.2 |
XP_011539306.1
|
8, 9 |
670 |
|
XM_017000652.1 |
XP_016856141.1
|
9, 10 |
814 |
|
XM_017000653.1 |
XP_016856142.1
|
10, 11 |
934 |
|
XM_017000654.1 |
XP_016856143.1
|
8, 9 |
771 |
|
XM_017000655.1 |
XP_016856144.1
|
9, 10 |
927 |
|
XM_017000656.1 |
XP_016856145.1
|
8, 9 |
674 |
|
XM_017000657.1 |
XP_016856146.1
|
7, 8 |
666 |
|
XM_017000658.1 |
XP_016856147.1
|
7, 8 |
691 |
|
XM_017000659.1 |
XP_016856148.1
|
6, 7 |
550 |
|
GenBank mRNA |
AK289805.1 |
- |
8, 9 |
733 |
|
AK295745.1 |
- |
7, 8 |
630 |
|
AK298129.1 |
- |
7, 8 |
691 |
|
AK303664.1 |
- |
6, 7 |
550 |
|
BC029500.1 |
- |
8, 9 |
734 |
5272319 |
BC041667.1 |
- |
7, 8 |
563 |
5743906 |
BX648945.1 |
- |
8, 9 |
743 |
|
U81602.1 |
- |
3, 4 |
373 |
|
View Full Product Details