Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033214.2 | NP_149991.2 | 1 | 1429 | |
GenBank mRNA | AK292241.1 | - | 1 | 1439 | |
AK312768.1 | - | 1 | 1421 | ||
BC029820.1 | - | 1 | 1421 | 5169374 | |
BC048274.1 | - | 1 | 1428 | 5166566 | |
BC058888.1 | - | 1 | 1419 | 5172337 | |
X78712.1 | - | 1 | 1429 |