Gene Transcripts

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
Chromosome Location
Chr. 9: 128504692 - 128542301 on Build GRCh38
UniGene ID
Hs.522418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003722.1 NP_001003722.1 12 1830
NM_001499.2 NP_001490.1 12 1830
XM_006717059.2 XP_006717122.1 14 1969
XM_006717060.2 XP_006717123.1 13 1942
XM_011518549.1 XP_011516851.1 14 2006
XM_011518550.1 XP_011516852.1 14 1857
XM_011518551.1 XP_011516853.1 13 1857
GenBank mRNA AF058922.1 - 12 1803
AK001767.1 - 7 1080
AK074801.1 - 11 1586
AK222842.1 - 12 1776
AK222854.1 - 12 1804
AK290533.1 - 11 1567
AL117584.1 - 10 1313
BC030012.1 - 12 1823 4829102

View Full Product Details