Gene Transcripts
- Gene Symbol
- GNB2
- Entrez Gene ID
- 2783
- Gene Name
- G protein subunit beta 2
- Gene Aliases
- Chromosome Location
- Chr. 7: 100673740 - 100679169 on Build GRCh38
- UniGene ID
- Hs.185172
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005273.3 |
NP_005264.2
|
3, 4 |
361 |
|
GenBank mRNA |
AF064853.1 |
- |
3, 4 |
361 |
|
AF501883.1 |
- |
2, 3 |
88 |
|
AK291592.1 |
- |
3, 4 |
236 |
|
AL535249.3 |
- |
3, 4 |
355 |
|
BC010073.2 |
- |
3, 4 |
285 |
4300639 |
BC012348.1 |
- |
3, 4 |
346 |
4556119 |
BC068003.1 |
- |
3, 4 |
271 |
6055863 |
BM763069.1 |
- |
3, 4 |
358 |
|
CR541729.1 |
- |
2, 3 |
88 |
|
CV569909.1 |
- |
3, 4 |
237 |
|
M16538.1 |
- |
3, 4 |
235 |
|
M36429.1 |
- |
2, 3 |
145 |
|
View Full Product Details