Gene Transcripts

Gene Symbol
GPR32
Entrez Gene ID
2854
Gene Name
G protein-coupled receptor 32
Gene Aliases
RVDR1
Chromosome Location
Chr. 19: 50770464 - 50771732 on Build GRCh38
UniGene ID
Hs.515555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001506.2 NP_001497.1 1 285
GenBank mRNA BC067453.1 - 1 148
BC067454.1 - 1 148 6971744
BC095544.1 - 1 286 6971954

View Full Product Details