Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 9 | 1107 | |
XM_005257253.1 | XP_005257310.1 | 9 | 1152 | ||
GenBank mRNA | AF055008.1 | - | 9 | 913 | |
AK000607.1 | - | 9 | 891 | ||
AK023348.1 | - | 6 | 395 | ||
AK222522.1 | - | 9 | 891 | ||
AK296090.1 | - | 9 | 845 | ||
AY124489.1 | - | 8 | 869 | ||
BC000324.2 | - | 9 | 1070 | 2821810 | |
BC010577.2 | - | 8 | 861 | 3457813 | |
BT006844.1 | - | 8 | 851 | ||
M75161.1 | - | 8 | 863 | ||
X62320.1 | - | 9 | 891 |