Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000849.4 | NP_000840.2 | 3 | 392 | |
GenBank mRNA | AB209816.1 | - | 2 | 101 | |
AK313444.1 | - | 2 | 110 | ||
BC000088.2 | - | 2 | 97 | 3509784 | |
BC008790.2 | - | 3 | 392 | 3630923 | |
BE392471.1 | - | 3 | 372 | ||
BT019945.1 | - | 2 | 82 | ||
CR542108.1 | - | 2 | 82 | ||
EF363096.1 | - | 2 | 99 | ||
J05459.1 | - | 2 | 99 |