Gene Transcripts

Gene Symbol
HLA-DMA
Entrez Gene ID
3108
Gene Name
major histocompatibility complex, class II, DM alpha
Gene Aliases
D6S222E, DMA, HLADM, RING6
Chromosome Location
Chr. 6: 32948614 - 32953122 on Build GRCh38
UniGene ID
Hs.728759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006120.3 NP_006111.2 2 193
GenBank mRNA AK291858.1 - 2 129
AK298081.1 - 3 142
BC011447.1 - 2 166 4245221
BC026279.1 - 2 122 4703588
CR450324.1 - 2 107
X62744.1 - 2 152

View Full Product Details