Gene Transcripts

Gene Symbol
HLA-DQA2
Entrez Gene ID
3118
Gene Name
major histocompatibility complex, class II, DQ alpha 2
Gene Aliases
DX-ALPHA, HLA-DXA
Chromosome Location
Chr. 6: 32741386 - 32746887 on Build GRCh38
UniGene ID
Hs.591798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020056.4 NP_064440.1 2 349
GenBank mRNA M95727.1 - 1 189
M95728.1 - 1 189

View Full Product Details