Gene Transcripts

Gene Symbol
HOXB5
Entrez Gene ID
3215
Gene Name
homeobox B5
Gene Aliases
HHO.C10, HOX2, HOX2A, HU-1, Hox2.1
Chromosome Location
Chr. 17: 48591257 - 48593741 on Build GRCh38
UniGene ID
Hs.654456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002147.3 NP_002138.1 1 115
GenBank mRNA AK314964.1 - 1 172
BC117247.1 - 1 76 40125798
BQ073272.1 - 49
M92299.1 - 1 331

View Full Product Details