Gene Transcripts

Gene Symbol
HSPA6
Entrez Gene ID
3310
Gene Name
heat shock protein family A (Hsp70) member 6
Gene Aliases
HSP70B'
Chromosome Location
Chr. 1: 161524540 - 161526897 on Build GRCh38
UniGene ID
Hs.654614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002155.4 NP_002146.2 1 1008
GenBank mRNA AK093925.1 - 1 1008
AK223362.1 - 1 1008
AK295212.1 - 2 931
AK312756.1 - 1 1008
BC035665.1 - 1 998 5723718

View Full Product Details