Gene Transcripts

Gene Symbol
HTR1D
Entrez Gene ID
3352
Gene Name
5-hydroxytryptamine receptor 1D
Gene Aliases
5-HT1D, HT1DA, HTR1DA, HTRL, RDC4
Chromosome Location
Chr. 1: 23191895 - 23194729 on Build GRCh38
UniGene ID
Hs.121482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000864.4 NP_000855.1 1 822
GenBank mRNA AF498979.1 - 1 312
BC007720.2 - 1 822 4299633
BT007027.1 - 1 312
M81589.1 - 1 355

View Full Product Details