Gene Transcripts

Gene Symbol
IFIT1
Entrez Gene ID
3434
Gene Name
interferon induced protein with tetratricopeptide repeats 1
Gene Aliases
C56, G10P1, IFI-56, IFI-56K, IFI56, IFIT-1, IFNAI1, ISG56, P56, RNM561
Chromosome Location
Chr. 10: 89392546 - 89406487 on Build GRCh38
UniGene ID
Hs.20315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270927.1 NP_001257856.1 3 570
NM_001270928.1 NP_001257857.1 3 587
NM_001270929.1 NP_001257858.1 3 582
NM_001270930.1 NP_001257859.1 3 708
NM_001548.4 NP_001539.3 2 473
GenBank mRNA AK092813.1 - 2 1501
AK308688.1 - 3 474
AK314588.1 - 2 396
AL548182.3 - 2 376
AL711085.1 - 3 483
BC007091.1 - 2 398 4250452
BI670242.1 - 2 481
BT006667.1 - 1 306
CD106320.1 - 2 349
DA822484.1 - 2 455
DB003897.1 - 3 507
DB180094.1 - 3 517
X03557.1 - 2 370

View Full Product Details