Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145349.1 | NP_001138821.1 | 9 | 1892 | |
NM_005897.2 | NP_005888.1 | 9 | 1892 | ||
XM_005270839.4 | XP_005270896.1 | 9 | 1780 | ||
XM_006710623.3 | XP_006710686.1 | 8 | 1671 | ||
GenBank mRNA | AF156857.1 | - | 8 | 1675 | |
AK023880.1 | - | 9 | 1728 | ||
BC032544.1 | - | 9 | 1733 | 5535760 |