Gene Transcripts

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
Chromosome Location
Chr. 3: 122421901 - 122514939 on Build GRCh38
UniGene ID
Hs.161008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002264.3 NP_002255.3 13 1491
NR_026698.1 14 1614
XM_005247437.3 XP_005247494.1 13 1373
XM_005247439.3 XP_005247496.1 11 2121
XM_011512795.2 XP_011511097.1 14 1425
XM_011512797.2 XP_011511099.1 14 3088
XM_017006363.1 XP_016861852.1 14 3062
XM_017006364.1 XP_016861853.1 14 1491
XM_017006365.1 XP_016861854.1 9 1755
GenBank mRNA AK125626.1 - 10 1441
AK128280.1 - 11 2121
AK299030.1 - 13 1500
AK312713.1 - 14 1609
BC002374.2 - 13 1480 2822859
BC003009.1 - 13 1480 2822859
BC090864.1 - 13 1462 6173605
BT006959.1 - 12 1303
CR456743.1 - 12 1303
S75295.1 - 13 1349
U20620.1 - 11 1155

View Full Product Details