Gene Transcripts

Gene Symbol
MAGEB4
Entrez Gene ID
4115
Gene Name
MAGE family member B4
Gene Aliases
CT3.6
Chromosome Location
Chr. X: 30241940 - 30244193 on Build GRCh38
UniGene ID
Hs.113291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002367.3 NP_002358.1 1 1184
GenBank mRNA AK313769.1 - 1 1124
BC032852.2 - 1 1072 5267183
CR541779.1 - 1 988

View Full Product Details