Gene Transcripts
- Gene Symbol
- MUT
- Entrez Gene ID
- 4594
- Gene Name
- methylmalonyl-CoA mutase
- Gene Aliases
- MCM
- Chromosome Location
- Chr. 6: 49430360 - 49463328 on Build GRCh38
- UniGene ID
- Hs.485527
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000255.3 |
NP_000246.2
|
5, 6 |
1342 |
|
XM_005249143.3 |
XP_005249200.1
|
5, 6 |
1247 |
|
GenBank mRNA |
AK292568.1 |
- |
5, 6 |
1318 |
|
AK312611.1 |
- |
5, 6 |
1317 |
|
BC016282.1 |
- |
5, 6 |
1205 |
3908548 |
BT007434.1 |
- |
4, 5 |
1077 |
|
BX647789.1 |
- |
5, 6 |
1167 |
|
KF030882.1 |
- |
4, 5 |
1077 |
|
KU178201.1 |
- |
4, 5 |
1077 |
|
KU178202.1 |
- |
4, 5 |
1077 |
|
M65131.1 |
- |
5, 6 |
1153 |
|
View Full Product Details