Gene Transcripts

Gene Symbol
NDUFA8
Entrez Gene ID
4702
Gene Name
NADH:ubiquinone oxidoreductase subunit A8
Gene Aliases
CI-19KD, CI-PGIV, PGIV
Chromosome Location
Chr. 9: 122132466 - 122159819 on Build GRCh38
UniGene ID
Hs.495039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318195.1 NP_001305124.1 2 331
NM_014222.2 NP_055037.1 2 331
GenBank mRNA AF044953.1 - 2 256
AK301520.1 - 2 299
AK314135.1 - 2 291
BC001016.2 - 2 235 3345138
BU602971.1 - 2 333

View Full Product Details