Gene Transcripts

Gene Symbol
PABPC3
Entrez Gene ID
5042
Gene Name
poly(A) binding protein cytoplasmic 3
Gene Aliases
PABP3, PABPL3, tPABP
Chromosome Location
Chr. 13: 25096138 - 25098567 on Build GRCh38
UniGene ID
Hs.458280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030979.2 NP_112241.2 1 411
GenBank mRNA AF132026.1 - 1 373
AK313336.1 - 1 393
AL136900.1 - 1 394
BC027617.2 - 1 377 4829121
BC045608.1 - 1 387 5273503
BG718483.1 - 1 416

View Full Product Details