Gene Transcripts
- Gene Symbol
- PHKB
- Entrez Gene ID
- 5257
- Gene Name
- phosphorylase kinase regulatory subunit beta
- Gene Aliases
- Chromosome Location
- Chr. 16: 47461131 - 47701523 on Build GRCh38
- UniGene ID
- Hs.78060
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000293.2 |
NP_000284.1
|
8, 9 |
836 |
|
NM_001031835.2 |
NP_001027005.1
|
9, 10 |
948 |
|
XM_005255983.4 |
XP_005256040.1
|
8, 9 |
799 |
|
XM_005255984.4 |
XP_005256041.1
|
9, 10 |
1099 |
|
XM_017023282.1 |
XP_016878771.1
|
2, 3 |
93 |
|
XM_017023283.1 |
XP_016878772.1
|
9, 10 |
921 |
|
XM_017023284.1 |
XP_016878773.1
|
8, 9 |
802 |
|
GenBank mRNA |
AK309226.1 |
- |
8, 9 |
799 |
|
BC033657.1 |
- |
9, 10 |
927 |
3913693 |
CR936710.1 |
- |
9, 10 |
982 |
|
X84908.1 |
- |
8, 9 |
808 |
|
View Full Product Details