Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007106.3 | NP_009037.1 | 3 | 1305 | |
GenBank mRNA | AF044221.1 | - | 3 | 264 | |
AK056580.1 | - | 3 | 1200 | ||
AK307361.1 | - | 3 | 249 | ||
AK309253.1 | - | 3 | 234 | ||
AK311846.1 | - | 3 | 348 | ||
AL080177.1 | - | 3 | 269 | ||
BC044582.1 | - | 3 | 748 | 5262176 | |
BC059385.1 | - | 3 | 645 | 5287541 | |
CR533458.1 | - | 3 | 160 |