Gene Transcripts
- Gene Symbol
- PPP2R1A
- Entrez Gene ID
- 5518
- Gene Name
- protein phosphatase 2 scaffold subunit Aalpha
- Gene Aliases
- MRD36, PP2A-Aalpha, PP2AAALPHA, PR65A
- Chromosome Location
- Chr. 19: 52189802 - 52226425 on Build GRCh38
- UniGene ID
- Hs.467192
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014225.5 |
NP_055040.2
|
11, 12 |
1649 |
|
NR_033500.1 |
|
10, 11 |
1548 |
|
XM_017026929.1 |
XP_016882418.1
|
11, 12 |
1878 |
|
GenBank mRNA |
AK090488.1 |
- |
11, 12 |
1883 |
|
AK091387.1 |
- |
11, 12 |
1253 |
|
AK130000.1 |
- |
10, 11 |
1157 |
|
AK290593.1 |
- |
11, 12 |
1437 |
|
AK291932.1 |
- |
11, 12 |
1432 |
|
AK293174.1 |
- |
12, 13 |
1400 |
|
AK293493.1 |
- |
10, 11 |
1273 |
|
AK299012.1 |
- |
11, 12 |
1304 |
|
AK315700.1 |
- |
11, 12 |
1465 |
|
BC001537.2 |
- |
11, 12 |
1395 |
2987938 |
BC026733.1 |
- |
11, 12 |
1412 |
|
CR450340.1 |
- |
11, 12 |
1354 |
|
J02902.1 |
- |
11, 12 |
1498 |
|
View Full Product Details