Gene Transcripts

Gene Symbol
PRODH
Entrez Gene ID
5625
Gene Name
proline dehydrogenase 1
Gene Aliases
HSPOX2, PIG6, POX, PRODH1, PRODH2, TP53I6
Chromosome Location
Chr. 22: 18912774 - 18936553 on Build GRCh38
UniGene ID
Hs.517352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195226.1 NP_001182155.1 4 435
NM_016335.4 NP_057419.4 5 850
GenBank mRNA AB209472.1 - 4 501
BC036534.1 - 4 659
BC068260.1 - 4 651
BC094736.1 - 4 480
BC118597.1 - 4 435 40108132
BC121809.1 - 4 435 40108133
KJ534920.1 - 4 646
U79754.1 - 5 850

Gene Symbol
LOC102724788
Entrez Gene ID
102724788
Gene Name
proline dehydrogenase 1, mitochondrial
Gene Aliases
Chromosome Location
Chr. 22|NT_187389: 138086 - 161619 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006724935.3 XP_006724998.1 4 679
XM_006724936.3 XP_006724999.1 4 654
XM_017030166.1 XP_016885655.1 4 776
XM_017030167.1 XP_016885656.1 4 1385
XM_017030168.1 XP_016885657.1 4 497
XM_017030169.1 XP_016885658.1 3 1120
XM_017030170.1 XP_016885659.1 3 406

View Full Product Details