Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204402.1 | NP_001191331.1 | 2 | 346 | |
NM_002764.3 | NP_002755.1 | 5 | 754 | ||
GenBank mRNA | AK223169.1 | - | 5 | 571 | |
AK297968.1 | - | 6 | 592 | ||
AK307748.1 | - | 4 | 570 | ||
AK312706.1 | - | 5 | 668 | ||
AK316467.1 | - | 2 | 260 | ||
BC001605.1 | - | 5 | 649 | 3542584 | |
D00860.1 | - | 5 | 671 | ||
X15331.1 | - | 5 | 615 |