Gene Transcripts

Gene Symbol
PSMD13
Entrez Gene ID
5719
Gene Name
proteasome 26S subunit, non-ATPase 13
Gene Aliases
HSPC027, Rpn9, S11, p40.5
Chromosome Location
Chr. 11: 236675 - 252984 on Build GRCh38
UniGene ID
Hs.134688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002817.3 NP_002808.3 7 742
NM_175932.2 NP_787128.2 5 748
XM_011520235.2 XP_011518537.1 7 875
GenBank mRNA AB009398.1 - 7 569
AF083245.1 - 7 554
AF086708.1 - 7 500
AF107837.1 - 7 533
AK094775.1 - 5 579
AK222895.1 - 7 573
AK295944.1 - 6 539
AK307447.1 - 6 467
AK314733.1 - 7 1004
BC001100.2 - 7 555 3506530
BC001747.1 - 7 523 3535807
BM456586.1 - 7 573
BT007307.1 - 7 500
BX647314.1 - 6 477

View Full Product Details