Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002889.3 | NP_002880.1 | 3 | 327 | |
XM_017012491.1 | XP_016867980.1 | 3 | 338 | ||
GenBank mRNA | AB015632.1 | - | 3 | 282 | |
AK092804.1 | - | 1 | 1494 | ||
AK312197.1 | - | 3 | 281 | ||
BC000069.2 | - | 3 | 258 | 3506067 | |
BM919890.1 | - | 2 | 262 | ||
BX448753.2 | - | 3 | 327 | ||
CR541992.1 | - | 2 | 210 | ||
CR542026.1 | - | 2 | 210 | ||
U77594.2 | - | 3 | 270 |