Gene Transcripts

Gene Symbol
RP1
Entrez Gene ID
6101
Gene Name
retinitis pigmentosa 1 (autosomal dominant)
Gene Aliases
DCDC4A, ORP1
Chromosome Location
Chr. 8: 54509448 - 54630834 on Build GRCh38
UniGene ID
Hs.732820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006269.1 NP_006260.1 4 1727
XM_017013721.1 XP_016869210.1 9 2890
XM_017013722.1 XP_016869211.1 4 1743
GenBank mRNA AF141021.1 - 4 1727
AF146592.1 - 4 1675

View Full Product Details