Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005983.3 | NP_005974.2 | 3 | 576 | |
NM_032637.3 | NP_116026.1 | 3 | 576 | ||
XM_011514081.2 | XP_011512383.1 | 3 | 630 | ||
XM_011514082.2 | XP_011512384.1 | 3 | 630 | ||
GenBank mRNA | AB050979.1 | - | 2 | 356 | |
AB050980.1 | - | 2 | 356 | ||
AB050981.1 | - | 2 | 356 | ||
AK291255.1 | - | 3 | 508 | ||
AK295152.1 | - | 3 | 450 | ||
AY029177.1 | - | 3 | 408 | ||
BC001441.2 | - | 3 | 495 | 3138709 | |
BC007441.2 | - | 3 | 499 | 2962938 | |
U33761.1 | - | 3 | 539 |