Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003058.3 | NP_003049.2 | 2 | 615 | |
GenBank mRNA | AB075951.1 | - | 2 | 619 | |
AK290787.1 | - | 2 | 519 | ||
BC030978.1 | - | 2 | 615 | ||
BC039899.1 | - | 2 | 496 | 5186229 | |
X98333.1 | - | 2 | 585 |