Gene Transcripts
- Gene Symbol
- STK11
- Entrez Gene ID
- 6794
- Gene Name
- serine/threonine kinase 11
- Gene Aliases
- LKB1, PJS, hLKB1
- Chromosome Location
- Chr. 19: 1205799 - 1228435 on Build GRCh38
- UniGene ID
- Hs.515005
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000455.4 |
NP_000446.1
|
3, 4 |
1584 |
|
XM_005259617.2 |
XP_005259674.1
|
3, 4 |
1094 |
|
XM_005259618.3 |
XP_005259675.1
|
3, 4 |
1094 |
|
XM_011528209.1 |
XP_011526511.1
|
4, 5 |
1225 |
|
GenBank mRNA |
AB209553.1 |
- |
3, 4 |
559 |
|
AF035625.1 |
- |
3, 4 |
807 |
|
AF217978.1 |
- |
4, 5 |
1430 |
|
AK314858.1 |
- |
3, 4 |
595 |
|
BC007981.2 |
- |
3, 4 |
919 |
3689780 |
BC019334.1 |
- |
3, 4 |
740 |
3946005 |
KU178339.1 |
- |
3, 4 |
469 |
|
U63333.1 |
- |
3, 4 |
469 |
|
View Full Product Details