Gene Transcripts

Gene Symbol
SULT2A1
Entrez Gene ID
6822
Gene Name
sulfotransferase family 2A member 1
Gene Aliases
DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa
Chromosome Location
Chr. 19: 47870466 - 47886397 on Build GRCh38
UniGene ID
Hs.515835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003167.3 NP_003158.2 1 190
GenBank mRNA AK289380.1 - 1 132
AK313415.1 - 1 117
BC020755.1 - 1 107 4733628
BG563083.1 - 1 170
BX509490.1 - 1 191
L02337.1 - 1 115
L20000.1 - 1 102
S43859.1 - 1 115
U08024.1 - 1 70
U08025.1 - 1 114
X70222.1 - 1 102
X84816.1 - 1 95

View Full Product Details