Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003200.3 | NP_003191.1 | 18 | 1870 | |
XM_006722852.3 | XP_006722915.1 | 18 | 2045 | ||
XM_006722853.3 | XP_006722916.1 | 18 | 2045 | ||
XM_011528215.2 | XP_011526517.1 | 17 | 3343 | ||
XM_011528216.2 | XP_011526518.1 | 18 | 2044 | ||
XM_011528217.2 | XP_011526519.1 | 18 | 2047 | ||
XM_011528219.2 | XP_011526521.1 | 18 | 2042 | ||
XM_011528223.2 | XP_011526525.1 | 18 | 1953 | ||
XM_017027177.1 | XP_016882666.1 | 18 | 2044 | ||
XM_017027180.1 | XP_016882669.1 | 15 | 1971 | ||
XM_017027181.1 | XP_016882670.1 | 18 | 1956 | ||
XM_017027183.1 | XP_016882672.1 | 18 | 2128 | ||
XM_017027184.1 | XP_016882673.1 | 18 | 2048 | ||
GenBank mRNA | BC011665.2 | - | 15 | 1614 | |
BC110580.1 | - | 14 | 1841 | 40007779 | |
M24405.1 | - | 9 | 1159 | ||
M31222.1 | - | 1870 | |||
M31523.1 | - | 17 | 1831 |