Gene Transcripts
- Gene Symbol
- TIAL1
- Entrez Gene ID
- 7073
- Gene Name
- TIA1 cytotoxic granule-associated RNA binding protein-like 1
- Gene Aliases
- TCBP, TIAR
- Chromosome Location
- Chr. 10: 119573465 - 119597247 on Build GRCh38
- UniGene ID
- Hs.501203
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033925.1 |
NP_001029097.1
|
8, 9 |
1262 |
|
NM_001323964.1 |
NP_001310893.1
|
9, 10 |
1587 |
|
NM_001323965.1 |
NP_001310894.1
|
9, 10 |
1330 |
|
NM_001323967.1 |
NP_001310896.1
|
9, 10 |
1279 |
|
NM_001323968.1 |
NP_001310897.1
|
9, 10 |
1324 |
|
NM_001323969.1 |
NP_001310898.1
|
9, 10 |
1273 |
|
NM_001323970.1 |
NP_001310899.1
|
9, 10 |
1324 |
|
NM_003252.3 |
NP_003243.1
|
8, 9 |
1211 |
|
NR_136661.1 |
|
8, 9 |
1211 |
|
NR_136662.1 |
|
9, 10 |
1330 |
|
NR_136663.1 |
|
9, 10 |
1273 |
|
XM_017016606.1 |
XP_016872095.1
|
5, 6 |
1215 |
|
GenBank mRNA |
AB209260.1 |
- |
4, 5 |
2021 |
|
AK290695.1 |
- |
8, 9 |
1164 |
|
AK290984.1 |
- |
8, 9 |
861 |
|
AK291239.1 |
- |
8, 9 |
844 |
|
AK295243.1 |
- |
9, 10 |
724 |
|
AL833106.1 |
- |
8, 9 |
1147 |
|
AY633610.1 |
- |
8, 9 |
655 |
|
BC030025.1 |
- |
9, 10 |
1592 |
4821759 |
D64015.1 |
- |
8, 9 |
687 |
|
M96954.1 |
- |
8, 9 |
692 |
|
View Full Product Details