Gene Transcripts

Gene Symbol
TTK
Entrez Gene ID
7272
Gene Name
TTK protein kinase
Gene Aliases
CT96, ESK, MPH1, MPS1, MPS1L1, PYT
Chromosome Location
Chr. 6: 80004147 - 80043577 on Build GRCh38
UniGene ID
Hs.169840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166691.1 NP_001160163.1 18 2229
NM_003318.4 NP_003309.2 18 2220
XM_011536099.2 XP_011534401.1 18 2194
XM_011536100.2 XP_011534402.1 18 2217
XM_017011242.1 XP_016866731.1 19 2749
GenBank mRNA AK292460.1 - 18 2192
AK315696.1 - 18 2137
AX775733.1 - 18 2192
BC000633.2 - 18 2183 3343925
BC032858.2 - 18 2185 5270012
X70500.1 - 5 642

View Full Product Details